missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyrazine-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10691416
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5521-55-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00068241
InChI Key RBYJWCRKFLGNDB-UHFFFAOYSA-N
Synonym 5-methyl-2-pyrazinecarboxylic acid, 2-methylpyrazine-5-carboxylic acid, pyrazinecarboxylic acid, 5-methyl, unii-b861rs5nhi, 5-methylpyrazinecarboxylic acid, 5-methyl pyrazine-2-carboxylic acid, 5-methyl-pyrazine-2-carboxylic acid, 2-carboxy-5-methylpyrazine, 2-methyl-5-pyrazinoic acid, b861rs5nhi
PubChem CID 122831
IUPAC Name 5-methylpyrazine-2-carboxylic acid
SMILES CC1=NC=C(N=C1)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 138.13
Percent Purity 97%
Chemical Name or Material 5-Methylpyrazine-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'