missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyluridine, 99%, Thermo Scientific Chemicals
£46.20 - £131.00
Chemical Identifiers
CAS | 1463-10-1 |
---|---|
Molecular Formula | C10H14N2O6 |
Molecular Weight (g/mol) | 258.23 |
MDL Number | MFCD00006535 |
InChI Key | DWRXFEITVBNRMK-JXOAFFINSA-N |
Synonym | 5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine |
PubChem CID | 445408 |
ChEBI | CHEBI:45996 |
IUPAC Name | 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
SMILES | CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15461369
|
Thermo Scientific Alfa Aesar
H64259.14 |
25 g |
£46.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15471369
|
Thermo Scientific Alfa Aesar
H64259.22 |
100 g |
£131.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1463-10-1 | |
258.23 | |
DWRXFEITVBNRMK-JXOAFFINSA-N | |
445408 | |
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
C10H14N2O6 | |
MFCD00006535 | |
5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine | |
CHEBI:45996 | |
CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O |
Specifications
1463-10-1 | |
White | |
MFCD00006535 | |
5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine | |
CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O | |
258.23 | |
CHEBI:45996 | |
99% | |
5-Methyluridine |
183°C to 184°C | |
C10H14N2O6 | |
90164 | |
DWRXFEITVBNRMK-JXOAFFINSA-N | |
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione | |
445408 | |
258.23 | |
Solid |
Safety and Handling
missing translation for 'einecsNumber' : 215-973-9
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only