missing translation for 'onlineSavingsMsg'
Learn More

5-(morpholinomethyl)isoxazole-3-carbaldehyde, Thermo Scientific™

Product Code. 10637580
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 893749-85-4
Molecular Formula C9H12N2O3
Molecular Weight (g/mol) 196.206
InChI Key AGLGFPIEVWZSLU-UHFFFAOYSA-N
Synonym 5-morpholinomethyl isoxazole-3-carbaldehyde, 5-morpholin-4-ylmethyl-1,2-oxazole-3-carbaldehyde, 5-morpholin-4-ylmethyl isoxazole-3-carbaldehyde, 5-morpholin-4-yl methyl-1,2-oxazole-3-carbaldehyde
PubChem CID 20099059
IUPAC Name 5-(morpholin-4-ylmethyl)-1,2-oxazole-3-carbaldehyde
SMILES C1COCCN1CC2=CC(=NO2)C=O

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 196.21
Percent Purity 97%
Chemical Name or Material 5-(morpholinomethyl)isoxazole-3-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.