Learn More
5-Nitro-m-xylene, 99%, Thermo Scientific Chemicals
£168.00
Chemical Identifiers
CAS | 99-12-7 |
---|---|
Molecular Formula | C8H9NO2 |
Molecular Weight (g/mol) | 151.17 |
MDL Number | MFCD00007269 |
InChI Key | BYFNZOKBMZKTSC-UHFFFAOYSA-N |
Synonym | 5-nitro-m-xylene, 3,5-dimethylnitrobenzene, 3,5-dimethyl-1-nitrobenzene, benzene, 1,3-dimethyl-5-nitro, nitroxylol, m-xylene, 5-nitro, unii-5hvw2qtf0d, ccris 3121, 5hvw2qtf0d, 5-nitro-m-xylol |
PubChem CID | 7426 |
IUPAC Name | 1,3-dimethyl-5-nitrobenzene |
SMILES | CC1=CC(=CC(C)=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322047
|
Thermo Scientific Alfa Aesar
L07762.18 |
50 g | |||||||
Description
5-Nitro-m-xylene was used in aerobic oxidation of aromatic anilines to aromatic azo compounds using gold nanoparticles supported on TiO2 as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Nitro-m-xylene was used in aerobic oxidation of aromatic anilines to aromatic azo compounds using gold nanoparticles supported on TiO2 as a catalyst.
Solubility
Insoluble in water.
Notes
Incompatible materials are Strong oxidizing agents, Strong bases, Strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
99-12-7 | |
151.17 | |
BYFNZOKBMZKTSC-UHFFFAOYSA-N | |
7426 | |
CC1=CC(=CC(C)=C1)[N+]([O-])=O |
C8H9NO2 | |
MFCD00007269 | |
5-nitro-m-xylene, 3,5-dimethylnitrobenzene, 3,5-dimethyl-1-nitrobenzene, benzene, 1,3-dimethyl-5-nitro, nitroxylol, m-xylene, 5-nitro, unii-5hvw2qtf0d, ccris 3121, 5hvw2qtf0d, 5-nitro-m-xylol | |
1,3-dimethyl-5-nitrobenzene |
Specifications
99-12-7 | |
273°C | |
MFCD00007269 | |
1936130 | |
Insoluble in water. | |
CC1=CC(=CC(C)=C1)[N+]([O-])=O | |
151.17 | |
151.17 | |
5-Nitro-m-xylene |
69°C to 73°C | |
C8H9NO2 | |
UN3447 | |
5-nitro-m-xylene, 3,5-dimethylnitrobenzene, 3,5-dimethyl-1-nitrobenzene, benzene, 1,3-dimethyl-5-nitro, nitroxylol, m-xylene, 5-nitro, unii-5hvw2qtf0d, ccris 3121, 5hvw2qtf0d, 5-nitro-m-xylol | |
BYFNZOKBMZKTSC-UHFFFAOYSA-N | |
1,3-dimethyl-5-nitrobenzene | |
7426 | |
99% |
Safety and Handling
GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
H301+H311+H331-H332-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROXYLENES, SOLID
missing translation for 'einecsNumber' : 202-732-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only