missing translation for 'onlineSavingsMsg'
Learn More
Alfa Aesar™ 5-Nitroindoline, 97% Product Code.: 11407106

Alfa Aesar™ 5-Nitroindoline, 97%

Product Code. 11407106
25 g
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy

Product Code. 11407106

Brand: Alfa Aesar™ B25417.14

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 32692-19-6
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD00005709
InChI Key WJQWYAJTPPYORB-UHFFFAOYSA-N
Synonym 5-nitroindoline, 1h-indole, 2,3-dihydro-5-nitro, indoline, 5-nitro, 2,3-dihydro-5-nitro-1h-indole, ccris 3306, 5-nitro-2,3-dihydro-indole, 5-nitroindolin, 5-nitro indoline, pubchem7442, acmc-1aera
PubChem CID 36219
IUPAC Name 5-nitro-2,3-dihydro-1H-indole
SMILES C1CNC2=C1C=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 92°C to 94°C
Quantity 25 g
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 5-Nitroindoline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Alfa Aesar™ 5-Nitroindoline, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.