missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroisatin, 97%, Thermo Scientific Chemicals

Product Code. 10785401
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-09-6
Molecular Formula C8H4N2O4
Molecular Weight (g/mol) 192.13
MDL Number MFCD00005720
InChI Key UNMYHYODJHKLOC-UHFFFAOYSA-N
Synonym 5-nitroisatin, 5-nitroindoline-2,3-dione, 5-nitroindole-2,3-dione, 1h-indole-2,3-dione, 5-nitro, indole-2,3-dione, 5-nitro, isatin, 5-nitro, n-nitroisatin, ccris 4031, 2,3-dihydro-5-nitroindole-2,3-dione, 5-nitro-2,3-dihydro-1h-indole-2,3-dione
PubChem CID 4669250
IUPAC Name 5-nitro-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C(=O)C(=O)N2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 251°C
Color Orange to Orange-Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 456
Formula Weight 192.13
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroisatin, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Nitroisatin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'