missing translation for 'onlineSavingsMsg'
Learn More

5-Oxo-1-(2-thienylmethyl)pyrrolidine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. p-2702825
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10374382

Brand: Thermo Scientific Maybridge DFP00358DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175136-92-2
Molecular Formula C10H11NO3S
Molecular Weight (g/mol) 225.262
MDL Number MFCD00084927
InChI Key HNEDKCTVOYJYQI-UHFFFAOYSA-N
PubChem CID 614397
IUPAC Name 5-oxo-1-(thiophen-2-ylmethyl)pyrrolidine-3-carboxylic acid
SMILES C1C(CN(C1=O)CC2=CC=CS2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-oxo-1-2-thienylmethyl pyrrolidine-3-carboxylic acid, 5-oxo-1-thiophen-2-ylmethyl pyrrolidine-3-carboxylic acid, 4-carboxy-1-then-2-yl pyrrolidin-2-one, 3-pyrrolidinecarboxylicacid, 5-oxo-1-2-thienylmethyl, 5-oxo-1-thien-2-ylmethyl pyrrolidine-3-carboxylic acid, maybridge1_003341, 3-pyrrolidinecarboxylicacid,5-oxo-1-2-thienylmethyl, 5-oxo-1-2-thienylmethyl-3-pyrrolidinecarboxylic acid, 5-oxo-1-2-thienylmethyl-3-pyrrolidinecarboxylic acid #, 5-oxo-1-thiophen-2-ylmethyl-pyrrolidine-3-ca rboxylic acid
Formula Weight 225.27
Percent Purity 97%
Chemical Name or Material 5-Oxo-1-(2-thienylmethyl)pyrrolidine-3-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.