missing translation for 'onlineSavingsMsg'
Learn More

5-Oxo-5-(2-thienyl)valeric acid, 97%, Thermo Scientific™

Product Code. 11358076
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22971-62-6
Molecular Formula C9H10O3S
Molecular Weight (g/mol) 198.236
MDL Number MFCD00151848
InChI Key GSQMRVFOHNXKOR-UHFFFAOYSA-N
Synonym 5-oxo-5-2-thienyl valeric acid, 5-oxo-5-thiophen-2-yl pentanoic acid, 4-2-thienoyl butyric acid, 5-oxo-5-thien-2-ylpentanoic acid, 5-oxo-5-2-thienyl pentanoic acid, 2-thiophenepentanoicacid, d-oxo, acmc-1cj8s, 4-2-thenoyl butyric acid, 5-2-thienoyl butyric acid, 5-thien-2-yl-5-oxopentanoic acid
PubChem CID 255155
IUPAC Name 5-oxo-5-thiophen-2-ylpentanoic acid
SMILES C1=CSC(=C1)C(=O)CCCC(=O)O

Specifications

Melting Point 94°C to 97°C
Quantity 1 g
Beilstein 141775
Formula Weight 198.24
Percent Purity 97%
Chemical Name or Material 5-Oxo-5-(2-thienyl)valeric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'