missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10571682
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 99924-18-2
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00105447
InChI Key RUKDIKJSGDVSIF-UHFFFAOYSA-N
Synonym 5-phenyloxazole-4-carboxylic acid, 5-phenyl-4-oxazolecarboxylic acid, 4-oxazolecarboxylic acid, 5-phenyl, 5-phenyl-oxazole-4-carboxylic acid, buttpark 15357-35, 4-carboxy-5-phenyl-1,3-oxazole, 4-oxazolecarboxylicacid, 5-phenyl, 5-phenyl-1,3-oxazol-4-ylcarboxylic acid, saltdata: free, 2-amino-1,2,3,4-tetrahydro-5,8-dimethoxy-2-naphthalenecarboxylic acid methyl ester
PubChem CID 2776299
IUPAC Name 5-phenyl-1,3-oxazole-4-carboxylic acid
SMILES OC(=O)C1=C(OC=N1)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 189.17
Percent Purity 97%
Chemical Name or Material 5-Phenyl-1, 3-oxazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'