missing translation for 'onlineSavingsMsg'
Learn More

5-Phenylnicotinic acid, ≥95%, Thermo Scientific™

Product Code. 10503032
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 10177-12-5
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
MDL Number MFCD03086176
InChI Key VKFXHYRIHRTEIV-UHFFFAOYSA-N
Synonym 5-phenylnicotinic acid, 3-pyridinecarboxylic acid, 5-phenyl, 5-phenyl-3-pyridinecarboxylic acid, 3-phenyl-5-pyridinecarboxylic acid, 5-phenylnicotinicacid, akos bar-0484, acmc-1bstf, 3-carboxy-5-phenylpyridine, 5-phenylnicotinic acid, 95+%, 3-pyridinecarboxylicacid,5-phenyl
PubChem CID 346160
IUPAC Name 5-phenylpyridine-3-carboxylic acid
SMILES OC(=O)C1=CN=CC(=C1)C1=CC=CC=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 5-Phenylnicotinic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'