missing translation for 'onlineSavingsMsg'
Learn More

5-(Phenylsulfonyl)thiophene-2-sulfonyl chloride, tech. 90%, Thermo Scientific™

Product Code. 11312587
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166964-37-0
Molecular Formula C10H7ClO4S3
Molecular Weight (g/mol) 322.792
MDL Number MFCD00052143
InChI Key GSLVRDAYMHUGTF-UHFFFAOYSA-N
Synonym 5-phenylsulfonyl thiophene-2-sulfonyl chloride, 5-phenylsulfonyl thiophene-2-sulfonylchloride, 5-benzenesulfonyl thiophene-2-sulfonyl chloride, 2-benzenesulfonylthiophene-5-sulfonyl chloride, 5-phenylthiophene-2,5-disulphonyl chloride, pubchem5455, 5-benzenesulphonylthiophene-2-sulphonyl chloride, acmc-1c42s, 5-phenylsulphonyl thiophene-2-sulphonyl chloride, 2-phenylsulfonylthiophene-5-sulfonyl chloride
PubChem CID 580877
IUPAC Name 5-(benzenesulfonyl)thiophene-2-sulfonyl chloride
SMILES C1=CC=C(C=C1)S(=O)(=O)C2=CC=C(S2)S(=O)(=O)Cl

Specifications

Melting Point 102°C to 105°C
Quantity 250 mg
UN Number UN3261
Beilstein 7818655
Sensitivity Moisture sensitive
Formula Weight 322.81
Percent Purity 90%
Grade Technical
Chemical Name or Material 5-(Phenylsulfonyl)thiophene-2-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'