missing translation for 'onlineSavingsMsg'
Learn More

5-Pyridin-2-ylthiophene-2-carbaldehyde, Thermo Scientific™

Product Code. 10552832
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 132706-12-8
Molecular Formula C10H7NOS
Molecular Weight (g/mol) 189.232
InChI Key VAGVVFZWHTULBM-UHFFFAOYSA-N
Synonym 5-pyridin-2-yl thiophene-2-carbaldehyde, 5-2-pyridyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-2-pyridinyl, 5-pyridin-2-yl thiophene-2-carboxaldehyde, 5-2-pyridinyl-2-thiophenecarboxaldehyde, pubchem16361, acmc-1c1gm, 5-2-pyridinyl-2-thiophenecarbaldehyde, 5-2-pyridinyl-2-thiophenecarbaldehyde #, thiophene-2-carboxaldehyde, 5-2-pyridyl
PubChem CID 605127
IUPAC Name 5-pyridin-2-ylthiophene-2-carbaldehyde
SMILES C1=CC=NC(=C1)C2=CC=C(S2)C=O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 189.24
Percent Purity 97%
Chemical Name or Material 5-Pyridin-2-ylthiophene-2-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'