missing translation for 'onlineSavingsMsg'
Learn More

(5-Pyrrolidin-1-ylpyrid-2-yl)methanol, 97%, Thermo Scientific™

Product Code. 10295594
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10295594

Brand: Thermo Scientific Maybridge CC52909DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 930110-98-8
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.24
MDL Number MFCD09702371
InChI Key GLWULQJNDPJFDR-UHFFFAOYSA-N
Synonym 5-pyrrolidin-1-ylpyrid-2-yl methanol, 5-pyrrolidin-1-ylpyridin-2-yl methanol, 5-pyrrolidin-1-yl pyridin-2-yl methanol, 5-pyrrolidinyl-2-pyridyl methan-1-ol, 5-pyrrolidine-1yl-pyrid-2-yl methanol, 2-pyridinemethanol,5-1-pyrrolidinyl, 1-6-hydroxymethyl pyridin-3-yl pyrrolidine, 2-hydroxymethyl-5-pyrrolidin-1-yl pyridine
PubChem CID 24229532
SMILES OCC1=NC=C(C=C1)N1CCCC1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
IUPAC Name (5-pyrrolidin-1-ylpyridin-2-yl)methanol
Formula Weight 178.23
Percent Purity 95%
Chemical Name or Material (5-Pyrrolidin-1-ylpyrid-2-yl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.