Learn More
Thermo Scientific Chemicals 5-Sulfosalicylic acid dihydrate, 99+%
Brand: Thermo Scientific Chemicals 268792500
27.08 GBP valid until 2024-12-31
Use promo code "21665" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
5-Sulfosalicylic acid dihydrate | |
5965-83-3, 97-05-2 | |
100.0 | |
White | |
99+% | |
C9H14O6S | |
250 g | |
01,1118 | |
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd | |
NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
2-hydroxy-5-sulfobenzoic acid; bis(methane) | |
2723734 | |
99+% |
p.a. | |
99.0 | |
106.0°C to 110.0°C | |
Authentic | |
Plastic bottle | |
MFCD00007508,MFCD00149540 | |
11, 411 | |
15, 9094 | |
Solubility in water: 127g/L (20°C). Other solubilities: soluble in alcohol and ether | |
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O | |
250.27 | |
254.22 | |
Crystalline Powder |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only