missing translation for 'onlineSavingsMsg'
Learn More

5-(tert-Butyl)-3-(chloromethyl)-1,2,4-oxadiazole, 95+%, Thermo Scientific™

Product Code. 10173184
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-41-1
Molecular Formula C7H11ClN2O
Molecular Weight (g/mol) 174.628
InChI Key AIVSLDAXUIRICJ-UHFFFAOYSA-N
PubChem CID 2796738
IUPAC Name 5-tert-butyl-3-(chloromethyl)-1,2,4-oxadiazole
SMILES CC(C)(C)C1=NC(=NO1)CCl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 5-tert-butyl-3-chloromethyl-1,2,4-oxadiazole, 3-chloromethyl-5-1,1-dimethylethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-1,1-dimethylethyl, 3-chloromethyl-5-t-butyl-1,2,4-oxadiazole, 3-chloromethyl-5-tert-butyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl-5-1,1-dimethylethyl
Formula Weight 174.63
Percent Purity 95%
Chemical Name or Material 5-(tert-Butyl)-3-(chloromethyl)-1, 2, 4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'