missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(tert-Butyl)-3-(chloromethyl)-1,2,4-oxadiazole, 95+%, Thermo Scientific™
£12.75 - £222.00
Chemical Identifiers
CAS | 175205-41-1 |
---|---|
Molecular Formula | C7H11ClN2O |
Molecular Weight (g/mol) | 174.628 |
InChI Key | AIVSLDAXUIRICJ-UHFFFAOYSA-N |
PubChem CID | 2796738 |
IUPAC Name | 5-tert-butyl-3-(chloromethyl)-1,2,4-oxadiazole |
SMILES | CC(C)(C)C1=NC(=NO1)CCl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10602653
|
Thermo Scientific Maybridge
SEW00330CB |
250 mg | Amber Glass Bottle |
£12.75
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10099521
|
Thermo Scientific Maybridge
SEW00330DA |
1 g | Amber Glass Bottle |
£40.35
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10173184
|
Thermo Scientific Maybridge
SEW00330EA |
10 g | Amber Glass Bottle |
£222.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
175205-41-1 | |
174.628 | |
2796738 | |
CC(C)(C)C1=NC(=NO1)CCl |
C7H11ClN2O | |
AIVSLDAXUIRICJ-UHFFFAOYSA-N | |
5-tert-butyl-3-(chloromethyl)-1,2,4-oxadiazole |
Specifications
175205-41-1 | |
C7H11ClN2O | |
AIVSLDAXUIRICJ-UHFFFAOYSA-N | |
5-tert-butyl-3-(chloromethyl)-1,2,4-oxadiazole | |
2796738 | |
95% |
95% | |
5-tert-butyl-3-chloromethyl-1,2,4-oxadiazole, 3-chloromethyl-5-1,1-dimethylethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-1,1-dimethylethyl, 3-chloromethyl-5-t-butyl-1,2,4-oxadiazole, 3-chloromethyl-5-tert-butyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl-5-1,1-dimethylethyl | |
CC(C)(C)C1=NC(=NO1)CCl | |
174.628 | |
174.63 | |
5-(tert-Butyl)-3-(chloromethyl)-1, 2, 4-oxadiazole |