Learn More
5-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific Chemicals
£36.40 - £126.00
Chemical Identifiers
CAS | 131747-40-5 |
---|---|
Molecular Formula | C7H4F3NO2 |
Molecular Weight (g/mol) | 191.11 |
MDL Number | MFCD07774128 |
InChI Key | RKFDCELCLIZRRH-UHFFFAOYSA-N |
Synonym | 5-trifluoromethyl nicotinic acid, 5-trifluoromethyl pyridine-3-carboxylic acid, 5-trifluoromethylinicotinic acid, 5-trifluoromethyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylicacid, 5-trifluoromethyl, 5-trifluoromethy pyridine-3-carboxylic acid |
PubChem CID | 10976305 |
IUPAC Name | 5-(trifluoromethyl)pyridine-3-carboxylic acid |
SMILES | OC(=O)C1=CC(=CN=C1)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15450869
|
Thermo Scientific Alfa Aesar
H63906.MD |
250 mg | |||||||
15440869
|
Thermo Scientific Alfa Aesar
H63906.03 |
1 g |
£126.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
131747-40-5 | |
191.11 | |
RKFDCELCLIZRRH-UHFFFAOYSA-N | |
10976305 | |
OC(=O)C1=CC(=CN=C1)C(F)(F)F |
C7H4F3NO2 | |
MFCD07774128 | |
5-trifluoromethyl nicotinic acid, 5-trifluoromethyl pyridine-3-carboxylic acid, 5-trifluoromethylinicotinic acid, 5-trifluoromethyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylicacid, 5-trifluoromethyl, 5-trifluoromethy pyridine-3-carboxylic acid | |
5-(trifluoromethyl)pyridine-3-carboxylic acid |
Specifications
131747-40-5 | |
White | |
MFCD07774128 | |
5-trifluoromethyl nicotinic acid, 5-trifluoromethyl pyridine-3-carboxylic acid, 5-trifluoromethylinicotinic acid, 5-trifluoromethyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylicacid, 5-trifluoromethyl, 5-trifluoromethy pyridine-3-carboxylic acid | |
OC(=O)C1=CC(=CN=C1)C(F)(F)F | |
191.11 | |
191.11 | |
Solid |
184°C to 189°C | |
C7H4F3NO2 | |
UN2811 | |
RKFDCELCLIZRRH-UHFFFAOYSA-N | |
5-(trifluoromethyl)pyridine-3-carboxylic acid | |
10976305 | |
97% | |
5-(Trifluoromethyl)nicotinic acid |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only