Learn More
5-(Trifluoromethyl)pyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
£24.90 - £408.00
Chemical Identifiers
CAS | 80194-69-0 |
---|---|
Molecular Formula | C7H4F3NO2 |
Molecular Weight (g/mol) | 191.109 |
MDL Number | MFCD04113632 |
InChI Key | NJHGVAYLDHROPT-UHFFFAOYSA-N |
Synonym | 5-trifluoromethyl pyridine-2-carboxylic acid, 5-trifluoromethyl picolinic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl pyridine-2-carboxylicacid, pubchem15164, acmc-209pj2 |
PubChem CID | 10910349 |
IUPAC Name | 5-(trifluoromethyl)pyridine-2-carboxylic acid |
SMILES | C1=CC(=NC=C1C(F)(F)F)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15402369
|
Thermo Scientific Alfa Aesar
H64937.03 |
1 g |
£24.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15412369
|
Thermo Scientific Alfa Aesar
H64937.06 |
5 g |
£86.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15469266
|
Thermo Scientific Alfa Aesar
H64937.14 |
25 g |
£408.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-(Trifluoromethyl)pyridine-2-carboxylic acid is an intermediate used in the synthesis of β-secretase (BACE) inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-(Trifluoromethyl)pyridine-2-carboxylic acid is an intermediate used in the synthesis of β-secretase (BACE) inhibitors.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
80194-69-0 | |
191.109 | |
NJHGVAYLDHROPT-UHFFFAOYSA-N | |
10910349 | |
C1=CC(=NC=C1C(F)(F)F)C(=O)O |
C7H4F3NO2 | |
MFCD04113632 | |
5-trifluoromethyl pyridine-2-carboxylic acid, 5-trifluoromethyl picolinic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl pyridine-2-carboxylicacid, pubchem15164, acmc-209pj2 | |
5-(trifluoromethyl)pyridine-2-carboxylic acid |
Specifications
80194-69-0 | |
Brown | |
MFCD04113632 | |
Slightly soluble in water. | |
C1=CC(=NC=C1C(F)(F)F)C(=O)O | |
191.109 | |
191.11 | |
Powder |
133°C to 137°C | |
C7H4F3NO2 | |
5-trifluoromethyl pyridine-2-carboxylic acid, 5-trifluoromethyl picolinic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl pyridine-2-carboxylicacid, pubchem15164, acmc-209pj2 | |
NJHGVAYLDHROPT-UHFFFAOYSA-N | |
5-(trifluoromethyl)pyridine-2-carboxylic acid | |
10910349 | |
95% | |
5-(Trifluoromethyl)pyridine-2-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only