missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labcon 50 mL ViewPoint™ Centrifuge Thermochromic Tubes
Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C
Brand: Labcon 3220-320-006-9
39.67 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 9.07180kg
Description
- Real-time temperature signaling
- Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C
- With attached flat screw caps
- Black graduations and white writing surface
- ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Sperm Motility Assay (SMA) Mouse Embryo Assay (MEA) validated safe for cell culture
- Polypropylene USP Class VI resin
- Centrifuging to 12,500 x G
- Freezable to -80°C
- Made in USA
Specifications
ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity | |
Clear to Blue color change at 0°C to 4°C | |
Single-use | |
10/IntegraPack™ Bag | |
10/IntegraPack™ Bag | |
ViewPoint™ | |
Autoclavable | |
29 mm | |
Attached Flat Integral Elastomeric Seal Screw Cap | |
29 mm | |
Conical Centrifuge Tube |
Purple | |
50 mL | |
Polypropylene Resin | |
10 Tubes / IntegraPack™ bag/5 bags / Pk./10 Pk. / case | |
Non-sterile | |
Room Temperature | |
115 mm | |
Conical | |
Yes | |
5 to 45 in increments of 1 | |
12,500 x g |