Learn More
Agilent BioTek 50 TS Microplate Washer
GREENER_CHOICE
The BioTek 50 TS washer offers excellent microplate washing performance. For Research Use Only. Not for use in diagnostic procedures.
Brand:  Agilent BioTek 50TS12-SN
 View more versions of this product
Product Code. 17207789
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well
The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.
Services
12 months
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE and TUV marked. RoHS compliant.
Optional (2 to 4 L)
Programmable in minutes and seconds, up to 30 min.
9.8 kg
25 to 3000 μL/well
150 to 1000 μL/well/sec. (manifold dependent)
50TS12
4.3 in. Color LCD Touch Screen Display
USB (1)
24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold)
Microplate Washer
1 to 10
4.3 in. LCD Color Touch Screen
One positive displacement syringe drive
96-well plates only
22 lb.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Agilent BioTek 50 TS Microplate Washer > For 96-well washing; 12-well manifold; Without buffer switching