Learn More
Agilent BioTek 50 TS Microplate Washer

The BioTek 50 TS washer offers excellent microplate washing performance. For Research Use Only. Not for use in diagnostic procedures.
Brand: Agilent BioTek 50TS8F-SN
Description
The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well
The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.
Services
12 months

Specifications
CE and TUV marked. RoHS compliant. | |
Optional (2 to 4 L) | |
Programmable in minutes and seconds, up to 30 min. | |
9.8 kg | |
25 to 3000 μL/well | |
150 to 1000 μL/well/sec. (manifold dependent) | |
50TS8F | |
4.3 in. Color LCD Touch Screen Display |
USB (1) | |
24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold) | |
Microplate Washer | |
1 to 10 | |
4.3 in. LCD Color Touch Screen | |
One positive displacement syringe drive | |
96-well plates only; Vacuum Filtration | |
22 lb. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.