missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 50 TS Microplate Washer
GREENER_CHOICE

Product Code. 17267789 Shop All Agilent BioTek Products
50TS8M, Each
Click to view available options
Model:
50TS12
50TS12V
50TS16
50TS16V
50TS8
50TS8F
50TS8M
50TS8MF
50TS8MV
50TS8V
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17267789

Brand: Agilent BioTek 50TS8MSN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The BioTek 50 TS washer offers excellent microplate washing performance. For Research Use Only. Not for use in diagnostic procedures.

The BioTek 50 TS washer offers excellent microplate washing performance. Ideal for use as an ELISA washer, the system is also well suited for cell-based assay washing. The 50 TS can process biomagnetic or polystyrene bead and vacuum filtration protocols as well

The color touch screen provides a visual interface that makes creating protocols fast and intuitive. When used with the 800 TS absorbance reader or other detection system, the 50 TS delivers convenience and high-quality results to your laboratory plate washing workflows.

Warranty

12 months

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE and TUV marked. RoHS compliant.
Connections USB (1)
Bottles Optional (2 to 4 L)
Plate Type 24-, 96-, 384-well Plates (96- and 384-well: Standard and Low Profile Plates, Solid and Filter Bottom (Option), 24-well Plates with 4-well Manifold)
Soak Time Programmable in minutes and seconds, up to 30 min.
Type Microplate Washer
Weight (Metric) 9.8 kg
Wash Cycles 1 to 10
Wash Volume 25 to 3000 ÎĽL/well
Display 4.3 in. LCD Color Touch Screen
Flow Rate 150 to 1000 ÎĽL/well/sec. (manifold dependent)
Fluid Delivery Type One positive displacement syringe drive
Model 50TS8M
Includes 96-well plates only; Biomagnetic Separation (with available accessory module sold separately)
Automation Interface 4.3 in. Color LCD Touch Screen Display
Weight (English) 22 lb.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Agilent BioTek 50 TS Microplate Washer >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.