Learn More
500 ML ETHANOL 99.8+% (GLC) 0.7897G/ML ABSOLUTEduty will be charged at the current rate for
Brand: Fisher Chemical E/0650/08
182.69 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
500 ML ETHANOL 99.8+% (GLC) 0.7897G/ML ABSOLUTEduty will be charged at the current rate for
Code : 22
Additional Details : CAS Number : 64-17-5 Weight : 0.39500kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Safety and Handling
- Ethanol, absolute
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P337+P313-If eye irritation persists: Get medical advice/attention.