Learn More
Spectrum™ Labs Spectra/Por™ 7 25,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
Brand:  Spectrum™ 132127T
 View more versions of this product
Product Code. 15310712
  • £146.00 / 1m
Explore more special offers
Includes:
  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks
Description
Description
  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application
Specifications
Specifications
25 kDa
22 mm
3.7 mL/cm vol./length
1 m Tubing/Kit