Learn More
Spectrum™ Labs Spectra/Por™ 7 25,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
Brand:  Spectrum™ 132 125T
 View more versions of this product
Product Code. 15390702
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes:
  • 1meter of pre-treated tubing, a pair of closures and 5 opening picks
Description
Description
  • Hydrophilic, symmetric porositiy
  • Equilibrated in water
  • Packaged wet in preservative solution of 0.05% sodium azide
  • For small experiments or qualifying a membrane for a particular application
Specifications
Specifications
25 kDa
7.5 mm
0.45 mL/cm vol./length
1 m Tubing/Kit