missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits

With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

Brand:  Spectrum™ 132660T

Code : NEW

Additional Details : Weight : 0.18000kg

 View more versions of this product

Product Code. 15310752

  • £223.00 / 5m
Estimated Shipment: 12-09-2024
to see stock.

Includes:
  • 1m or 5m of Spectra/Por 1 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers

Description

Description

  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)

Specifications

Specifications

6 to 8 kDa
25.5 mm
5.1 mL/cm vol./length
5 m Tubing/Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers