Learn More
Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
Marca: Spectrum™ 132 670T1
Code : NEW
Dettagli aggiuntivi : Weight : 0.10000kg
Includes:
- 1m or 5m of Spectra/Por 1 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
Descrizione
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water)
Specifica
6 to 8 kDa | |
64 mm | |
32 mL/cm vol./length | |
1 m Tubing/Kit |