Learn More
Spectrum™ Labs Spectra/Por™ 2 12-14 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
Brand: Spectrum™ 132684T
Code : NEW
Additional Details : Weight : 0.20000kg
Includes:
- 1m or 5m of Spectra/Por 2 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
Descripción
- For higher permeability applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water)
Especificaciones
12 to 14 kDa | |
76 mm | |
46 mL/cm vol./length | |
5 m Tubing/Kit |