missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 2 12-14 kD MWCO Standard RC Dry Dialysis Kits

With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated

Brand:  Spectrum™ 132676T

Code : NEW

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15390752

  • £125.00 / 5m
Estimated Shipment: 18-10-2024
to see stock.

Includes:
  • 1m or 5m of Spectra/Por 2 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers

Description

Description

  • For higher permeability applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)

Specifications

Specifications

12 to 14 kDa
6.5 mm
0.32 mL/cm vol./length
5 m Tubing/Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers