missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 50 mL Nalgene Oak Ridge Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor

Accommodate 50mL Oak Ridge tubes in Thermo Scientific™ Fiberlite™ rotors.

Brand:  Thermo Scientific™ 010-0377

92.48 GBP valid until 2024-11-30
Use promo code "21681" to get your promotional price.


Product Code. 12737077

  • £118.00 / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Accommodate 50mL Nalgene Oak Ridge tubes in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ridge Tube Adapter
50 mL
50mL Nalgene Oak Ridge Tube Adapter
50 mL Nalgene Oak Ridge Tube
Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers