missing translation for 'onlineSavingsMsg'
Learn More
Learn More
53BP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534674
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: NT2D1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Regulator that plays a central role in regulation of apoptosis and cell growth via its interactions. Regulates TP53 by enhancing the DNA binding and transactivation function of TP53 on the promoters of proapoptotic genes in vivo. Inhibits the ability of APPBP1 to conjugate NEDD8 to CUL1, and thereby decreases APPBP1 ability to induce apoptosis. Impedes cell cycle progression at G2/M. Its apoptosis-stimulating activity is inhibited by its interaction with DDX42.Specifications
53BP2 | |
Polyclonal | |
Unconjugated | |
TP53BP2 | |
53BP2; Apoptosis-stimulating of p53 protein 2; apoptosis-stimulating protein of p53, 2; ASPP2; BBP; Bcl2-binding protein; P PPP1R13A; P53BP2; PPP1R13A; Renal carcinoma antigen NY-REN-51; TP53BP2; transformation related protein 53 binding protein 2; Trp53bp2; tumor protein p53 binding protein 2; tumor protein p53 binding protein, 2; tumor suppressor p53-binding protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7159 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.92 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q13625 | |
TP53BP2 | |
Recombinant fragment corresponding to a region within amino acids 580 and 775 of ASPP2 (Uniprot ID#Q13625) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |