Learn More
Jenway™ 550 portable pH meter supplied in carry case with epoxy combination pH electrode, ATC probe and batteries
550 portable pH meter supplied in carry case with epoxy combination pH electrode, ATC probe and batteries. A small, lightweight general purpose pH meter offering 2 point pH calibration and automatic buffer recognition. Simultaneously displays both the temperature compensated pH readings and temperature.
Brand: Jenway™ 00394-NO
208.72 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Description
- Simultaneously displays pH and temperature
- Automatic calibration
- Manual or Automatic Temperature Compensation (ATC)
- Auto shut off 10 minutes
- 5000 hours battery life
- 2 year warranty
Specifications
Carry case with epoxy pH electrode, ATC probe, buffer sachets, and batteries | |
±0.05 | |
0.01 | |
0°C to 100°C | |
IP-65 | |
130 g | |
0°C to 60°C | |
pH and Temperature Meter |
±0.5°C | |
0 to 14 | |
2 | |
0.1°C | |
128 x 56 x 25 mm | |
2 Years | |
0°C to 50°C | |
5000 hr. |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85% - Multiple Manufacturers
Your input is important to us. Please complete this form to provide feedback related to the content on this product.