Learn More
Jenway™ 570 portable pH, mV meter supplied in carry case with epoxy combination pH electrode, ATC probe and batteries
570 portable pH, mV meter supplied in carry case with epoxy combination pH electrode, ATC probe and batteries. Is a general purpose pH, mV and temperature meter with an integrated rubber seal to ensure that it’s completely waterproof. This model displays either temperature compensated pH readings or electrode potential and temperature.
Brand: Jenway™ 10088-55
340.10 GBP valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
- Simultaneously displays pH and temperature
- Automatic calibration
- Manual or Automatic Temperature Compensation (ATC)
- Auto shut off 10 minutes
- 2500 hours battery life
- 2 year warranty
Specifications
Carry case with epoxy pH electrode, ATC probe, buffer sachets, and batteries | |
±1 | |
-2 to 16 | |
-1000 to 1000 | |
2 | |
0.1°C | |
141 x 71 x 32 mm | |
2 Years | |
0°C to 100°C | |
2500 hr. |
±0.04°C | |
±0.02 | |
0.01 | |
1mV | |
-39.9°C to +149.9°C | |
IP-66/67 | |
230 g | |
0°C to 100°C | |
pH, mV, and Temperature Meter |
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
29Hours
0Minutes
56Seconds
57 -
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
0Hours
23Minutes
3Seconds
2
Your input is important to us. Please complete this form to provide feedback related to the content on this product.