Learn More
Eppendorf™ Rotor for Centrifuge 5810 / 5810 R
Eppendorf Rotor for Centrifuge 5810/5810 R - precision redefined. High-capacity centrifugation with varied rotor options, a range of capacities and speeds, and versatile buckets to ensure efficient sample processing. Ideal for diverse applications, guaranteeing accuracy and reproducibility.
Brand: Eppendorf™ 5820754001
This item is not returnable.
View return policy
Description
- Precision-engineered for optimal sample separation.
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
- Accessories (adaptors and buckets) further customize your centrifuge setup.
Specifications
S-4-104 - Includes plate buckets (x4) | |
4 x 750 mL | |
3,900 rpm | |
Plate buckets (x4) |
Rotor | |
3,214 x g | |
Centrifuge 5810 / 5810 R |