missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5H-Chromeno[2,3-b]pyridin-5-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 197.19 |
Percent Purity | 97% |
Chemical Name or Material | 5H-Chromeno[2, 3-b]pyridin-5-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'