missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 5mL Polystyrene Immuno-Tubes

Use for determining hormone levels.

Brand:  Greiner Bio-One 115070

 View more versions of this product

Product Code. 13087573

  • £192.00 / Pack of 2000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Round bottom
  • Dia. x Height: 12 x 75mm
  • Available with either medium or high binding capacities
  • With or without star at bottom
  • Star at the bottom of the tubes serve to increase the surface and thus make it possible to bind amounts of antigens or antibodies
TRUSTED_SUSTAINABILITY
Specifications

Specifications

With star at the bottom of tube
Polystyrene
Medium-binding
12 mm
Determining hormone levels
Immuno Tube
75 mm
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers