missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One 5mL Polystyrene Immuno-Tubes

Use for determining hormone levels.

£126.54

Specifications

Material Polystyrene
Product Type Immuno Tube
For Use With (Application) Determining hormone levels
Length (Metric) 75 mm
Diameter (Metric) Outer 12 mm
View More Specs

Products 4
Product Code Brand Surface Includes Price Quantity & Availability  
Product Code Brand Surface Includes Price Quantity & Availability  
13067573
View Documents
Greiner Bio-One
115001
Medium-binding Without Star Bottom
£126.54
Pack of 2000
Please to purchase this item. Need a web account? Register with us today!
13077573
View Documents
Greiner Bio-One
115061
High-binding Without Star Bottom
£126.54
Pack of 2000
Please to purchase this item. Need a web account? Register with us today!
13087573
View Documents
Greiner Bio-One
115070
Medium-binding Star Bottom
£126.54
Pack of 2000
Please to purchase this item. Need a web account? Register with us today!
13097573
View Documents
Greiner Bio-One
115071
High-binding Star Bottom
£126.54
Pack of 2000
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Round bottom
  • Dia. x Height: 12 x 75mm
  • Available with either medium or high binding capacities
  • With or without star at bottom
  • Star at the bottom of the tubes serve to increase the surface and thus make it possible to bind amounts of antigens or antibodies
Specifications

Specifications

Polystyrene
Determining hormone levels
12 mm
Immuno Tube
75 mm
5 mL
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Greiner Bio-One 5mL Polystyrene Immuno-Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.