missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biochrom Quartz Microvolume Cuvette
Ultra micro cuvette with 5 to 7 μL working volume. Quartz Microvolume Cuvette for use with Ultrospec 2100 Pro and GeneQuant 1300/100.
Brand: Biochrom 80-2103-68
This item is not returnable.
View return policy
Description
- 5 mm ultra micro cuvette
- 5 to 7 μL working volume
- For use with Ultrospec 2100 pro (requires 80-2106-06 ultra microvolume cell holder) and GeneQuant 1300/100
Specifications
Ultra-Micro Cell | |
UV Grade Silica | |
5 mm | |
48 mm | |
12.5 mm | |
Rectangle |
5 to 7 μL | |
10 mm | |
190 to 1100 nm | |
Spectrophotometer, Photometer | |
Spectrophotometric Measurements | |
12 mm |