missing translation for 'onlineSavingsMsg'
Learn More

(5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone, 98%, Thermo Scientific Chemicals

Product Code. 15477258
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112741-49-8
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00074955
InChI Key MRUKRSQUUNYOFK-UHFFFAOYNA-N
Synonym tert-butyl 2r,3s---6-oxo-2,3-diphenyl-4-morpholinecarboxylate, 2r,3s-tert-butyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate, 5s,6r-n-boc-5,6-diphenyl-2-morpholinone, tert-butyl 2r,3s-6-oxo-2,3-diphenylmorpholine-4-carboxylate, 2r,3s---n-boc-6-oxo-2,3-diphenylmorpholine, tert-butyl-2r,3s---6-oxo-2,3-diphenyl-4-morpholinecarboxylate, 4-morpholinecarboxylicacid, 6-oxo-2,3-diphenyl-, 1,1-dimethylethyl ester, 2r,3s, pubchem11707
PubChem CID 981230
IUPAC Name tert-butyl (2R,3S)-6-oxo-2,3-diphenylmorpholine-4-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 206°C (decomposition)
Quantity 5 g
Formula Weight 353.41
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (5R,6S)-(-)-4-Boc-5,6-diphenyl-2-morpholinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'