missing translation for 'onlineSavingsMsg'
Learn More
5Z-7-Oxozeaenol, Thermo Scientific Chemicals Product Code.: 15428099

5Z-7-Oxozeaenol, Thermo Scientific Chemicals

Product Code. 15428099
1 mg
Click to view available options
Quantity:
1 mg
5 mg
This item is not returnable. View return policy

Product Code. 15428099

Brand: Thermo Scientific Alfa Aesar J66651.M^

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66018-38-0
Molecular Formula C19H22O7
Molecular Weight (g/mol) 362.378
MDL Number MFCD12912700
InChI Key NEQZWEXWOFPKOT-UHFFFAOYSA-N
Synonym 8,9,16-trihydroxy-14-methoxy-3-methyl-4,8,9,10-tetrahydro-3h-2-benzoxacyclotetradecine-1,7-dione
PubChem CID 53442201
IUPAC Name 5,6,15-trihydroxy-17-methoxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(14),2,8,15,17-pentaene-7,13-dione
SMILES CC1CC=CC(=O)C(C(CC=CC2=CC(=CC(=C2C(=O)O1)O)OC)O)O

Specifications

Color White
Quantity 1 mg
UN Number UN2811
Solubility Information Soluble in DMSO
Formula Weight 362.37
Physical Form Solid
Chemical Name or Material 5Z-7-Oxozeaenol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ20K021 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog NumberJ66651.M^,J66651.#0
Lot NumberW29I006 Certificate TypeCertificate of Analysis Date18/10/2022 Catalog NumberJ66651.M^,J66651.#0
Lot NumberP10D053 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ66651.M^,J66651.#0
Lot NumberX26D032 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ66651.M^,J66651.#0
Lot NumberS11B002 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ66651.M^,J66651.#0

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5Z-7-Oxozeaenol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.