missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dihydro-4(5H)-benzofuranone, 98%, Thermo Scientific Chemicals

£85.80 - £304.00

Chemical Identifiers

CAS 16806-93-2
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00215511
InChI Key DXWQOYPYNPSVRL-UHFFFAOYSA-N
Synonym 6,7-dihydro-4 5h-benzofuranone, 6,7-dihydrobenzofuran-4 5h-one, 4,5,6,7-tetrahydro-1-benzofuran-4-one, 6,7-dihydro-1-benzofuran-4 5h-one, 4 5h-benzofuranone, 6,7-dihydro, pubchem14044, 6,7-dihydro-4 5h benzo-furanone, 6,7-dihydro-5h-benzofuran-4-one, 6?7-dihydro-4 5h-benzofuranone, 6,7-dihydro-4-5h-benzofuranone
PubChem CID 2733541
IUPAC Name 6,7-dihydro-5H-1-benzofuran-4-one
SMILES C1CC2=C(C=CO2)C(=O)C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10365271
View Documents
Thermo Scientific Acros
330080050
5 g Glass bottle
£85.80
5g
Please to purchase this item. Need a web account? Register with us today!
10738041
View Documents
Thermo Scientific Acros
330080250
25 g Glass bottle
£304.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

16806-93-2
136.15
DXWQOYPYNPSVRL-UHFFFAOYSA-N
2733541
C1CC2=C(C=CO2)C(=O)C1
C8H8O2
MFCD00215511
6,7-dihydro-4 5h-benzofuranone, 6,7-dihydrobenzofuran-4 5h-one, 4,5,6,7-tetrahydro-1-benzofuran-4-one, 6,7-dihydro-1-benzofuran-4 5h-one, 4 5h-benzofuranone, 6,7-dihydro, pubchem14044, 6,7-dihydro-4 5h benzo-furanone, 6,7-dihydro-5h-benzofuran-4-one, 6?7-dihydro-4 5h-benzofuranone, 6,7-dihydro-4-5h-benzofuranone
6,7-dihydro-5H-1-benzofuran-4-one

Specifications

16806-93-2
100.0
Yellow to Brown
>110°C
97.5% min. (GC)
1.5300 to 1.5320
17, V,9, 585
Solubility in water: soluble. Other solubilities: soluble in methylene chloride,ether
C1CC2=C(C=CO2)C(=O)C1
136.15
136.15
Liquid
97.5
30.0°C to 34.0°C
115.0°C to 118.0°C (16.0 mmHg)
Authentic
C8H8O2
MFCD00215511
6,7-dihydro-4 5h-benzofuranone, 6,7-dihydrobenzofuran-4 5h-one, 4,5,6,7-tetrahydro-1-benzofuran-4-one, 6,7-dihydro-1-benzofuran-4 5h-one, 4 5h-benzofuranone, 6,7-dihydro, pubchem14044, 6,7-dihydro-4 5h benzo-furanone, 6,7-dihydro-5h-benzofuran-4-one, 6?7-dihydro-4 5h-benzofuranone, 6,7-dihydro-4-5h-benzofuranone
DXWQOYPYNPSVRL-UHFFFAOYSA-N
6,7-dihydro-5H-1-benzofuran-4-one
2733541
98%
6, 7-Dihydro-4(5H)-benzofuranone, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only