missing translation for 'onlineSavingsMsg'
Learn More
6,7-Dimethoxy-3,4-dihydroisoquinoline, 97%, Thermo Scientific Chemicals Product Code.: 15401039

6,7-Dimethoxy-3,4-dihydroisoquinoline, 97%, Thermo Scientific Chemicals

Product Code. 15401039
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15401039

Brand: Thermo Scientific Alfa Aesar H64018.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3382-18-1
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00266748
InChI Key NSLJVQUDZCZJLK-UHFFFAOYSA-N
PubChem CID 30058
IUPAC Name 6,7-dimethoxy-3,4-dihydroisoquinoline
SMILES COC1=CC2=C(C=NCC2)C=C1OC

Specifications

Melting Point 39°C to 40°C
Color Yellow
Boiling Point 155°C to 161°C (1 mmHg)
Refractive Index 1.593
Quantity 5 g
Beilstein 138269
Synonym 6,7-dimethoxy-3,4-dihydro-isoquinoline, 3,4-dihydro-6,7-dimethoxyisoquinoline, isoquinoline, 3,4-dihydro-6,7-dimethoxy, pubchem12828, ksc571e8h, 3,4-dihydro-6,7-dimethoxy-isoquinoline, isoquinoline,3,4-dihydro-6,7-dimethoxy, isoquinoline, 3,4-dihydro-6,7-dimethoxy-, hydrochloride 1:1
Formula Weight 191.23
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 6,7-Dimethoxy-3,4-dihydroisoquinoline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81500154 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberH64018.06,H64018.03,H64018.MD
Lot Number81400033 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH64018.06,H64018.03,H64018.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6,7-Dimethoxy-3,4-dihydroisoquinoline, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.