missing translation for 'onlineSavingsMsg'
Learn More
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride hydrate, 99+%, Thermo Scientific™ Product Code.: 10723251

6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride hydrate, 99+%, Thermo Scientific™

Product Code. 10723251
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10723251

Brand: Thermo Scientific Acros 265960050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 20232-39-7
Molecular Formula C11H14ClNO2
Molecular Weight (g/mol) 227.69
MDL Number MFCD11099370
InChI Key PQXVEYYRJHMTEV-UHFFFAOYSA-N
Synonym 6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, hcl, 6,7-dimethoxy-3,4-dihydroisoquinoline, chloride, 6,7-dimethoxy-3,4-dihydro-isoquinoline hydrochloride, pubchem17358, ksc496s5h, 3,4-dihydro-6,7-dimethoxyisoquinoline hydrochloride, 6,7-dimethoxy-3,4-dihydroisoquinoline hydro-chloride, 6,7-dimethoxy-3,4-dihydroisoquinolinehydrochloridehydrate
PubChem CID 2724664
SMILES Cl.COC1=CC2=C(C=NCC2)C=C1OC

Specifications

Melting Point 198.0°C to 202.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 5 g
IUPAC Name 6,7-dimethoxy-3,4-dihydroisoquinoline hydrochloride
Formula Weight 227.69
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material 6, 7-Dimethoxy-3, 4-dihydroisoquinoline hydrochloride hydrate
Show More Show Less
Product Identifier
  • 6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride hydrate, 99+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.