missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6,8-Dibromoimidazo[1,2-a]pyrazine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It is an Intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an Intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
It is an Intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
Specifications
Specifications
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 276.92 |
Percent Purity | 95% |
Chemical Name or Material | 6,8-Dibromoimidazo[1,2-a]pyrazine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'