Learn More
6,8-Dibromoimidazo[1,2-a]pyrazine, 95%, Thermo Scientific Chemicals
£56.70 - £208.00
Chemical Identifiers
CAS | 63744-22-9 |
---|---|
Molecular Formula | C6H3Br2N3 |
Molecular Weight (g/mol) | 276.92 |
MDL Number | MFCD08460056 |
InChI Key | UQCZZGIPIMJBCL-UHFFFAOYSA-N |
Synonym | 6,8-dibromoimidazo 1,2-a pyrazine, imidazo 1,2-a pyrazine, 6,8-dibromo, 6,8-dibromo-imidazo 1,2-a pyrazine, 6,8-dibromoimidazol 1,2-a pyrazine, pubchem14656, acmc-1b8pv, ksc351o9d, buttpark 180\02-82, 6,8 dibromoimidazo 1,2-a pyrazine, 6,8-dibomoimidazo 1,2-a pyrazine |
PubChem CID | 15025843 |
IUPAC Name | 6,8-dibromoimidazo[1,2-a]pyrazine |
SMILES | BrC1=CN2C=CN=C2C(Br)=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435378
|
Thermo Scientific Alfa Aesar
H59151.03 |
1 g |
£56.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15445378
|
Thermo Scientific Alfa Aesar
H59151.06 |
5 g |
£208.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
It is an Intermediate in organic syntheses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an Intermediate in organic syntheses.
Solubility
Slightly soluble in water.
Notes
Recommended storage temperature: 2 - 4°C. Incompatible with oxidizing agents.
Chemische Identifikatoren
63744-22-9 | |
276.92 | |
UQCZZGIPIMJBCL-UHFFFAOYSA-N | |
15025843 | |
BrC1=CN2C=CN=C2C(Br)=N1 |
C6H3Br2N3 | |
MFCD08460056 | |
6,8-dibromoimidazo 1,2-a pyrazine, imidazo 1,2-a pyrazine, 6,8-dibromo, 6,8-dibromo-imidazo 1,2-a pyrazine, 6,8-dibromoimidazol 1,2-a pyrazine, pubchem14656, acmc-1b8pv, ksc351o9d, buttpark 180\02-82, 6,8 dibromoimidazo 1,2-a pyrazine, 6,8-dibomoimidazo 1,2-a pyrazine | |
6,8-dibromoimidazo[1,2-a]pyrazine |
Spezifikation
63744-22-9 | |
MFCD08460056 | |
Slightly soluble in water. | |
BrC1=CN2C=CN=C2C(Br)=N1 | |
276.92 | |
276.92 | |
6,8-Dibromoimidazo[1,2-a]pyrazine |
C6H3Br2N3 | |
6,8-dibromoimidazo 1,2-a pyrazine, imidazo 1,2-a pyrazine, 6,8-dibromo, 6,8-dibromo-imidazo 1,2-a pyrazine, 6,8-dibromoimidazol 1,2-a pyrazine, pubchem14656, acmc-1b8pv, ksc351o9d, buttpark 180\02-82, 6,8 dibromoimidazo 1,2-a pyrazine, 6,8-dibomoimidazo 1,2-a pyrazine | |
UQCZZGIPIMJBCL-UHFFFAOYSA-N | |
6,8-dibromoimidazo[1,2-a]pyrazine | |
15025843 | |
95% |
Sicherheit und Handhabung
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only