missing translation for 'onlineSavingsMsg'
Learn More

6,8-Dichloro-2-oxo-2H-chromene-3-carboxylic acid, ≥95%, Thermo Scientific™

Product Code. 10354702
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 2199-86-2
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95+%
Packaging Amber Glass Bottle
Molecular Formula C10H4Cl2O4
MDL Number MFCD00047641
Quantity 250 mg
Synonym 6,8-dichloro-2-oxo-2h-chromene-3-carboxylic acid, 2h-1-benzopyran-3-carboxylicacid, 6,8-dichloro-2-oxo, 2h-1-benzopyran-3-carboxylic acid, 6,8-dichloro-2-oxo, opera_id_158, maybridge1_000531, 6,8-dichlorocoumarin-3-carboxylic acid, 2-oxo-6,8-dichloro-2h-1-benzopyran-3-carboxylic acid
InChI Key WJEHZKIVHXRVSR-UHFFFAOYSA-N
SMILES C1=C2C=C(C(=O)OC2=C(C=C1Cl)Cl)C(=O)O
IUPAC Name 6,8-dichloro-2-oxochromene-3-carboxylic acid
Molecular Weight (g/mol) 259.038
PubChem CID 657910
Formula Weight 259.04
Percent Purity 95%
Chemical Name or Material 6, 8-Dichloro-2-oxo-2H-chromene-3-carboxylic acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'