missing translation for 'onlineSavingsMsg'
Learn More
(6-Amino-3-pyridinyl)methanol, 97%, Thermo Scientific™ Product Code.: 10603835

(6-Amino-3-pyridinyl)methanol, 97%, Thermo Scientific™

Product Code. 10603835
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 10603835

Brand: Thermo Scientific Maybridge MO07396DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 113293-71-3
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
InChI Key TXPRFSOGPYITOT-UHFFFAOYSA-N
PubChem CID 2794829
IUPAC Name (6-aminopyridin-3-yl)methanol
SMILES C1=CC(=NC=C1CO)N

Specifications

Assay Percent Range 97%
Quantity 5 g
Synonym 6-aminopyridin-3-yl methanol, 6-amino-3-pyridinyl methanol, 6-amino-3-pyridinemethanol, 2-amino-5-hydroxymethylpyridine, 6-amino-pyridin-3-yl-methanol, 2-amino-5-hydroxymethyl pyridine, 2-amino-5-pyridinemethanol, 3-pyridinemethanol, 6-amino, 3-pyridinemethanol,6-amino, 6-amino-3-hydroxymethylpyridine
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material (6-Amino-3-pyridinyl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(6-Amino-3-pyridinyl)methanol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.