missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-7-bromo-9-methyl-7-deazapurine, 97%, Thermo Scientific Chemicals

Product Code. 15466338
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in the preparation of indoline derivatives as PERK inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of indoline derivatives as PERK inhibitors.

Solubility
Soluble in dichloromethane.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1337532-51-0
Molecular Formula C7H7BrN4
Molecular Weight (g/mol) 227.065
MDL Number MFCD20486185
InChI Key SCHJLAFNBDGWJN-UHFFFAOYSA-N
Synonym 5-bromo-7-methyl-7h-pyrrolo 2,3-d pyrimidin-4-amine, 5-bromo-7-methylpyrrolo 2,3-d pyrimidin-4-amine, 6-amino-7-bromo-9-methyl-7-deazapurine, 5-bromo-7-methyl-7h-pyrrolo-2,3-d pyrimidin-4-amine, 7h-pyrrolo 2,3-d pyrimidin-4-amine,5-bromo-7-methyl, 7h-pyrrolo 2,3-d pyrimidin-4-amine, 5-bromo-7-methyl
PubChem CID 66560938
IUPAC Name 5-bromo-7-methylpyrrolo[2,3-d]pyrimidin-4-amine
SMILES CN1C=C(C2=C1N=CN=C2N)Br

Specifications

Quantity 250 mg
Beilstein 21981671
Solubility Information Soluble in dichloromethane.
Formula Weight 227.06
Percent Purity 97%
Chemical Name or Material 6-Amino-7-bromo-9-methyl-7-deazapurine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.