missing translation for 'onlineSavingsMsg'
Learn More

6-Aminoquinoline, 98%, Thermo Scientific Chemicals

Product Code. 15117422
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 580-15-4
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
InChI Key RJSRSRITMWVIQT-UHFFFAOYSA-N
Synonym 6-aminoquinoline, 6-quinolinamine, quinoline, 6-amino, quinolin-6-ylamine, 6-quinolylamine, ccris 1681, 6 aminoquinoline, 3kqp, 6-amino quinoline, quinoline-6-amine
PubChem CID 11373
IUPAC Name quinolin-6-amine
SMILES C1=CC2=C(C=CC(=C2)N)N=C1

Specifications

Melting Point 115.0°C to 119.0°C
Boiling Point 192.0°C to 195.0°C (14.0 mmHg)
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 22, V,10, 303
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol, benzene, diethyl ether,heptane
Formula Weight 144.18
Percent Purity 98%
Chemical Name or Material 6-Aminoquinoline
compliance-icons
Product Identifier
  • 6-Aminoquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'