missing translation for 'onlineSavingsMsg'
Learn More
6-Azabicyclo[3.2.0]heptan-7-one, 98%, Thermo Scientific™ Product Code.: 10577031

6-Azabicyclo[3.2.0]heptan-7-one, 98%, Thermo Scientific™

Product Code. 10577031
500 mg, Glass bottle
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10577031

Brand: Thermo Scientific Acros 335835000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 22031-52-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 45.0°C to 50.0°C
Color White to Yellow
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H9NO
MDL Number MFCD01863277
Quantity 500 mg
Synonym 6-azabicyclo 3.2.0 heptan-7-one, 6-azabicyclo 3,2,0 heptan-7-one, 6-azabicyclo 3.2.0 heptane-7-one, 6-azabicyclo 3.2.0 heptan-7-one #
Solubility Information Solubility in water: .. Other solubilities: soluble in ether, chloroform, methanol
InChI Key AXDBIGNYXNSBHV-CRCLSJGQSA-N
SMILES O=C1N[C@@H]2CCC[C@H]12
IUPAC Name 6-azabicyclo[3.2.0]heptan-7-one
Molecular Weight (g/mol) 111.14
PubChem CID 549320
Formula Weight 111.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 6-Azabicyclo[3.2.0]heptan-7-one, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0301701 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number
Product Identifier
  • 6-Azabicyclo¢3.2.0!heptan-7-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Azabicyclo[3.2.0]heptan-7-one, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.